missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Grancalcin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119159
This item is not returnable.
View return policy
Description
Grancalcin Polyclonal Antibody for Western Blot, IHC (P)
This gene product, grancalcin, is a calcium-binding protein abundant in neutrophils and macrophages. It belongs to the penta-EF-hand subfamily of proteins which includes sorcin, calpain, and ALG-2. Grancalcin localization is dependent upon calcium and magnesium. In the absence of divalent cation, grancalcin localizes to the cytosolic fraction; with magnesium alone, it partitions with the granule fraction; and in the presence of magnesium and calcium, it associates with both the granule and membrane fractions, suggesting a role for grancalcin in granule-membrane fusion and degranulation.
Specifications
| Grancalcin | |
| Polyclonal | |
| Unconjugated | |
| Gca | |
| 5133401E04Rik; AI573844; Gca; GCL; grancalcin; grancalcin, EF-hand calcium binding protein; grancalcin, EF-hand calcium-binding protein; grancalcin, penta-EF-hand protein; null | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 227960, 25801, 295647 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P28676, Q8VC88 | |
| Gca | |
| Recombinant protein Grancalcin. The antigen corresponds to amino acid range 1-139 of the target protein. | |
| 200 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction