missing translation for 'onlineSavingsMsg'
Learn More

Gramine, 99%

Product Code. 10209833 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
10g
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10209833 500 g Glass bottle 500g
10366510 10 g Glass bottle 10g
10520941 25 g Glass bottle 25g
10162530 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10209833 Supplier Thermo Scientific Acros Supplier No. 120185000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 87-52-5 | C11H14N2 | 174.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-52-5
Molecular Formula C11H14N2
Molecular Weight (g/mol) 174.25
MDL Number MFCD00005629
InChI Key OCDGBSUVYYVKQZ-UHFFFAOYSA-N
Synonym gramine, 3-dimethylaminomethyl indole, donaxine, donaxin, gramin, 1h-indole-3-methanamine, n,n-dimethyl, beta-dimethylaminomethylindole, 1-1h-indol-3-yl-n,n-dimethylmethanamine, indol-3-ylmethyldimethylamine, 1h-indol-3-ylmethyl dimethylamine
PubChem CID 6890
ChEBI CHEBI:28948
IUPAC Name 1-(1H-indol-3-yl)-N,N-dimethylmethanamine
SMILES CN(C)CC1=CNC2=CC=CC=C21

Specifications

Melting Point 131.0°C to 139.0°C
Color Beige
Flash Point 167°C
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 500 g
Merck Index 15, 4569
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in alcohol,ether,toluene and chloroform, slightly soluble in cold acetone, practically insoluble in petr ether
Formula Weight 174.25
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Gramine
Show More Show Less
compliance-icons
Product Identifier
  • Gramine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.