missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Gramine

Product Code. 16434777
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16434777 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16434777 Supplier MedChemExpress Supplier No. HYN0166
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Gramine (Donaxine) is a natural alkaloid isolated from giant reed, acts as an active adiponectin receptor (AdipoR) agonist, with IC50s of 3.2 and 4.2 μM for AdipoR2 and AdipoR1, respectively. Gramine is also a human and mouse β2-Adrenergic receptor (β2-AR) agonist. Gramine (Donaxine) has anti-tumor, anti-viral and anti-inflammatory properties.

Specifications

Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 174.24 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-programmed cell death
CAS 87-52-5
Product Type Natural Products
Purity 99.39%
Solubility DMSO : ≥ 125 mg/mL (717.40 mM)
Synonym Donaxine
Target Adiponectin Receptor∣Adrenergic Receptor∣Reverse Transcriptase
Quantity 50 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.