missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRAMD4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24299-1-AP-20UL
This item is not returnable.
View return policy
Description
GRAMD4 also termed as Death inducing protein is a 578 amino-acid protein, which contains 1 GRAM domain. GRAMD4 as a multi-pass membrane protein localizes in the Mitochondrion membrane. Overexpression of GRAMD4 results in a strong apoptotic response involving caspase-3 activation and cleavage of poly (ADP-ribose)-polymerase. GRAMD4 is expressed in lung and in primary lung squamous cell carcinoma (LSCC) and shows up-regulation in mitochondria by E2F-1 after addition of 4-hydroxytamoxifen. This evidence suggests that GRAMD4 may be a potential target for cancer therapies.Specifications
| GRAMD4 | |
| Polyclonal | |
| Unconjugated | |
| GRAMD4 | |
| dA59H18.1, Death inducing protein, DIP, GRAM domain containing 4, GRAMD4, KIAA0767 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23151 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.59 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q6IC98 | |
| GRAMD4 | |
| GRAMD4 Fusion Protein Ag19140 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction