missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRAMD4 Mouse anti-Human, Clone: PCRP-GRAMD4-1A10, Novus Biologicals™
Mouse Monoclonal Antibody
£197.00 - £396.00
Specifications
| Antigen | GRAMD4 |
|---|---|
| Clone | PCRP-GRAMD4-1A10 |
| Concentration | 0.2 mg/ml |
| Dilution | Flow Cytometry 1-2 ug/million cells, Immunofluorescence 1-2 ug/ml, Protein Array |
| Applications | Flow Cytometry, Immunofluorescence, Peptide Array |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18387622
|
Novus Biologicals
NBP3-13894-20UG |
20 μg |
£197.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18366806
|
Novus Biologicals
NBP3-13894-100UG |
100 μg |
£396.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GRAMD4 Monoclonal antibody specifically detects GRAMD4 in Human samples. It is validated for Flow Cytometry, Immunofluorescence, Protein ArraySpecifications
| GRAMD4 | |
| 0.2 mg/ml | |
| Flow Cytometry, Immunofluorescence, Peptide Array | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q6IC98 | |
| 23151 | |
| IgG1 | |
| Protein A or G purified |
| PCRP-GRAMD4-1A10 | |
| Flow Cytometry 1-2 ug/million cells, Immunofluorescence 1-2 ug/ml, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| GRAM domain containing 4, GRAM domain-containing protein 4, KIAA0767 | |
| Recombinant full-length human GRAMD4 protein (Uniprot: Q6IC98) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title