missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GRAMD3 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA519380
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine, canine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 4,000.
Specifications
| GRAMD3 | |
| Polyclonal | |
| Unconjugated | |
| GRAMD2B | |
| 9030613F08Rik; 9130427A09Rik; AW061306; FLJ21313; GRAM domain containing 3; GRAM domain-containing protein 2B; GRAM domain-containing protein 3; GRAMD2B; GRAMD3; HCV NS3-transactivated protein 2; hepatitis C virus nonstructural protein 3-transactivating protein 2; NS3TP2 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 107022, 65983 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q6PEM6, Q96HH9 | |
| GRAMD2B, Gramd3 | |
| Synthetic peptide sequence (QTPESENSRD) corresponding to the internal amino acids of GRAMD3 (aa 274-283). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction