missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GRAMD2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569331
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 86%; Rat: 93%.
Participates in the organization of endoplasmic reticulum-plasma membrane contact sites (EPCS) with pleiotropic functions including STIM1 recruitment and calcium homeostasis. Constitutive tether that co-localize with ESYT2/3 tethers at endoplasmic reticulum-plasma membrane contact sites in a phosphatidylinositol lipid-dependent manner. Pre-marks the subset of phosphtidylinositol 4,5-biphosphate (PI(4,5)P2)-enriched EPCS destined for the store operated calcium entry pathway (SOCE).
Specifications
| GRAMD2 | |
| Polyclonal | |
| Unconjugated | |
| GRAMD2A | |
| BC064463; GRAM domain containing 2; GRAM domain containing 2A; GRAM domain-containing protein 2; GRAM domain-containing protein 2; GRAM domain-containing protein 2A; GRAM domain-containing protein 2A; GRAMD2; GRAMD2A; RGD1564007 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 196996 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8IUY3 | |
| GRAMD2A | |
| synthetic peptide directed towards the middle region of human GRAMD2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction