missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pro-Lab Diagnostics™ Gram Staining Kit
Description
- Pro-Lab Diagnostics offers an extensive range of ready-to-use and concentrated microbiological staining solutions
- Solutions come in a selection of convenient and economical sizes.
- All stains are manufactured in accordance with all current regulatory requirements
- CE labelled
Specifications
Specifications
| Certifications/Compliance | CE marked (98/78/EC) |
| Includes | Crystal Violet 250mL, Grams Iodine 250mL, Grams Differentiator 250mL and Neutral Red 250mL |
| Format | Liquid |
| For Use With (Application) | Gram’s Staining Method for the Initial Differentiation of Gram Positive and Gram Negative Bacteria |
| Stain Type | Gram |
| Volume (Metric) | 250 x 4 mL |
| Product Type | Gram Stain Kit |
Safety and Handling
Product Identifier
- Gram Staining Kit/Crystal Violet/Gram's Iodine/Gram's Differentiator/Neutral Red
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 2
- Specific target organ toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 3
- Flammable liquid Category 3
- H225-Highly flammable liquid and vapour.
- H332-Harmful if inhaled.
- H319-Causes serious eye irritation.
- H371-May cause damage to organs.
- H336-May cause drowsiness or dizziness.
- H412-Harmful to aquatic life with long lasting effects.
- H226-Flammable liquid and vapour.
- P273-Avoid release to the environment.
- P501-Dispose of contents/container to .
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P242-Use only non-sparking tools.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P271-Use only outdoors or in a well-ventilated area.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P370+P378-In case of fire: Use to extinguish.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P233-Keep container tightly closed.
- P405-Store locked up.
- MIXTURE LIST-Contain: ethanol, C.I. basicviolet 3, methanol,ethanol, acetone, ethanol, methanol, ethanol, methanol, glyc
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?