missing translation for 'onlineSavingsMsg'
Learn More

Pro-Lab Diagnostics™ Gram’s Differentiator

Product Code. 12906297
Click to view available options
Quantity:
2L
This item is not returnable. View return policy

Product Code. 12906297

Brand: Pro-Lab Diagnostics™ PL.7008

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use as a decolorizer in the Gram Staining Method for differentiating Gram positive and negative bacteria. Pro-Lab Diagnostics™ Gram’s Differentiator is an acetone solution manufactured to the highest standards, according to all current regulatory requirements, and CE labelled.

Pro-Lab Diagnostics™ Gram Stain solutions are manufactured in purpose-designed laboratories to the highest standards using the finest raw materials. Strict quality control and filtering techniques are employed prior to release to ensure that all solutions offer excellent performance characteristics.

  • Ready-to-use acetone solution in a convenient, economical size
  • Acts as a decolorizer to remove the initial stain from the cell wall of Gram-negative bacteria
  • Apply after staining with crystal violet and iodine solutions, and before using a counterstain

Specifications

Solution Type Ready-to-use Reagent
Type Gram's Differentiator - decolorization
Quantity 2L
Certifications/Compliance CE marked (98/78/EC)
Format Liquid
For Use With (Application) Gram’s Staining Method for the Initial Differentiation of Gram Positive and Gram Negative Bacteria
Color Clear, colorless
Boiling Point 56–78°C at 1013hPa
Composition Acetone 50 – <100%, Ethanol 25 – <50%, Methanol 2.5 – <5%
Storage Requirements Keep at temperature not exceeding 20°C
Stain Type Gram
Volume (Metric) 2 L
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Grams Differentiator
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H332-Harmful if inhaled.
  • H319-Causes serious eye irritation.
  • H371-May cause damage to organs.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
  • P501-Dispose of contents/container to .
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P370+P378-In case of fire: Use to extinguish.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contains : acetone, ethanol, methanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.