missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Gram Positive Bacteria LTA Monoclonal Antibody (G35C)
GREENER_CHOICE

Product Code. 11500542
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11500542 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11500542 Supplier Invitrogen™ Supplier No. MA17401

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The MA1-7401 antibody reacts with lipoteichoic acid from many gram positive bacteria. Reactive with Listeria monocytogenes (all serotypes), Streptococcus pneumoniae, Staphylococcus aureus, Staphylococcus epidermidis, Enterococcus faecium, Bacillus cereus, Bacillus subtilis, group B Streptococcus (weak). Non-reactive with Clostridium perfringens. Other species not tested. MA1-7401 has been successfully used in ELISA and Immunofluorescence applications. Suggested but not tested for Western blot and immunohistochemistry applications.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Gram Positive Bacteria LTA
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone G35C
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias Lipoteichoic acid
Host Species Mouse
Immunogen Lipoteichoic acid consistent with that of gram positive bacteria.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.