missing translation for 'onlineSavingsMsg'
Learn More

BD Gram Crystal Violet

Product Code. 11788193
Change view
Click to view available options
Quantity:
3.8 L
4 x 250mL
Unit Size:
1 L
3.8 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11788193 3.8 L 3.8 L
11778193 4 x 250mL 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11788193 Supplier BD Supplier No. 212526

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Can be used as a replacement reagent for the BD™ Gram Stain Kits. BD™ Gram Crystal Violet is used as the primary stain in a 4-step Gram staining process. Available in 250ml solution bottles for direct use or gallon containers for bulk storage.

Gram Stain reagents are used to stain microorganisms from cultures or specimens by the differential Gram method.

  • Complete stain kits reduce the mess associated with Gram staining procedures and include a compact tray for holding the 250mL reagent bottles
  • Break-resistant bottles features no-drip, keep-clean cap spouts that make regulating the flow easy and that help to prevent spots and spills
  • Color-coded labels allow quick identification of the reagent

Order Info

Available in packs of four 250mL bottles or individual one-gallon bottles

TRUSTED_SUSTAINABILITY

Specifications

Type Crystal Violet
Quantity 3.8 L
Certifications/Compliance CE
Format Liquid
For Use With (Application) Used to stain microorganisms from cultures or specimens by the differential gram method
Color Purple
Storage Requirements 15° to 30°C
Packaging Type Bulk storage container
Stain Type Crystal Violet
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.