missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ Grain Leather Drivers Gloves

Product Code. 11823490
Change view
Click to view available options
Size:
8
9
11
Unit Size:
10 pairs
Each
Product Code. Size unitSize
11823490 9 10 pairs
11813490 8 10 pairs
12681063 11 Each
3 options
This item is not returnable. View return policy

Product Code. 11823490

Brand: Honeywell™ 203019509

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Calf grain leather provides comfort and flexibility. The winged thumb and reinforced index finger make the glove more anatomical for additional ease of use.

  • Cowhide grain leather
  • Elasticated back
  • Gunn cut
  • Wing thumb
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE CATEGORY II Intermediary risks, 420-03 EEC Basic standards, EN 388-03 Mechanical hazards, EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Certificate Number N°0075/797/162/12/06/0358, EC Attestation Number N°0075/797/162/12/06/0358
Certifications/Compliance (Europe) CE Category: II Intermediary Risks, In accordance with : EN 420-03 EEC Basic Standards, and EN 388-16 Mechanical Hazards
Cut Style Gunn
Coating Material Leather
Color White
Cut Resistance Rating 1
Hand Specific
Material Grain Cowhide Leather
Product Type Driver Gloves
Size 9
Style Gunn
For Use With (Application) General Handling in Dry Environments
Glove Cuff Style Open
Includes Elasticized wrist
Lining Light and medium handling
Palm Type Leather
Quantity 10Pair/PK
Series 2030195
Tear Rating 2
Thumb Style Wing
Length (Metric) 25 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.