missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRAIL/RNF128 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97643PE
This item is not returnable.
View return policy
Description
GRAIL/RNF128 Polyclonal antibody specifically detects GRAIL/RNF128 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| GRAIL/RNF128 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| EC 6.3.2, EC 6.3.2.-, FLJ23516, Gene related to anergy in lymphocytes protein, ring finger protein 128GRAILE3 ubiquitin-protein ligase RNF128 | |
| Produced in rabbits immunized with E. coli-derived Human GRAIL/RNF128 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| PE | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 79589 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu