missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRAF Antibody (SR1016), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22504-100ul
This item is not returnable.
View return policy
Description
GRAF Monoclonal antibody specifically detects GRAF in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| GRAF | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| SR1016 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| FLJ42530, GRAFGTPase regulator associated with focal adhesion kinase pp125(FAK), KIAA0621rho GTPase-activating protein 26, Oligophrenin-1-like protein, OPHN1L1, OPHN1LGTPase regulator associated with focal adhesion kinase, Rho GTPase activating protein 26, Rho-type GTPase-activating protein 26 | |
| A synthesized peptide derived from human GRAF (Uniprot #: Q9UNA1) | |
| 100 μg | |
| Neuroscience, Stem Cell Markers | |
| 23092 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction