Learn More
Thermo Scientific™ Graduated Safelock Microcentrifuge Tubes
Product Code. 11920937 Shop All MBP Molecular BioProducts Products
Click to view available options
Sterility:
Non-sterile
Sterile
Capacity (Metric):
0.6 mL
1.5 mL
2 mL
Max. RCF:
24,000 x g
25,000 x g
30,000 x g
Packaging:
100 Tubes/Bag, 10 Bags/Unit, 10 Units/Cs.
1000/Pk. in 10 bags of 100
50 Tubes/Bag, 10 Bags/Unit, 10 Units/Cs.
500 Tubes/Bag, 5 Bags/Cs.
Unit Size:
Pack of 1000
Pack of 500
Product Code. 11920937
Brand: Thermo Scientific™ 3459IW
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Easily confirm your microcentrifuge tube is locked for superior security during centrifugation.
Find the security of a screw-cap tube with the ease-of-use of a snap-cap tube with Thermo Scientific™ Graduated Safelock Microcentrifuge Tubes. These tubes provide superior security during centrifugation. Simply position the hinged locking ball into the groove to prevent accidental cap opening, avoiding evaporation and ensuring sample integrity.
Safelock Lid Design
  • Simple one-hand operation snaps the hinged cap closed and slides the locking ball into place safeguarding valuable samples
  • Hinged locking ball reduces stress on the thumb
  • Smooth locking surface reduces the possibility of ripped or torn gloves
  • Graduation marks on the microcentrifuge tube allow for effective sample reference
Highest Quality
  • Manufactured from virgin polypropylene these microcentrifuge tubes ensure sample integrity and security during boiling, freezing, incubation and centrifugation
  • Exceptionally stable performance under the stress of centrifugation preventing tube breakage
  • Sterile product is certified free of RNase, DNase, DNA, ATP, bioburden and endotoxins
  • Nonsterile product is certified free of RNase, DNase, DNA and pyrogens
Packaging  Options
  • A variety of packaging options are available to help with contamination control in the lab
  • For stringent sterility requirements individually wrapped sterile tubes are available
Recommended For:
Daily and rigorous laboratory usage including centrifugation, boiling, incubation, freezing or sample storage.
TRUSTED_SUSTAINABILITY
Tekniske data
Certifications/Compliance Certified free of RNase, DNase, DNA and pyrogens
Capacity (Metric) 2 mL
Color Natural
Material Polypropylene
Packaging 500 Tubes/Bag, 5 Bags/Cs.
Sterility Non-sterile
Graduated Yes
Max. RCF 25,000 x g
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.