Learn More
Cytiva Grade GF/A Filter for Personal Air Samplers
These aerosol sampling and particulate monitoring filters provide high flow rates and minimal sample interference. Cytiva Whatman Grade GF/A card-mounted filters are used in static sample and personal air sampler applications.
Brand: Cytiva 18208296
125.68 GBP valid until 2024-12-31
Use promo code "23266" to get your promotional price.
Code : 69
Additional Details : Weight : 0.08550kg
Description
- Heat resistant (up to 500°C)
- Made of high-quality borosilicate glass fibers without added binders
- Low moisture affinity and high loading capacity
Specifications
Disc | |
For Air sampling | |
100 Pieces | |
32 mm |
Borosilicate Glass, Card Mount | |
GF/A (This data shown may be based on historic Whatman records. As is normal practice, customers should determine and validate for themselves (as necessary) that the products are suitable for their specific application.) | |
32 mm | |
32 mm |
Safety and Handling
- FILTER GLASS CIRCLE
Signal Word
- Warning
Hazard Category
- Carcinogenicity Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H351-Suspected of causing cancer.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : glass, oxide, chemicals
- UNKNOWN ACUTE-100 per cent of the mixture consists of component(s) of unknown acute toxicity.
- UNKNOWN AQUATIC-Contains 100% of components with unknown hazards to the aquatic environment.