missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Grade 600 Cellulose General Purpose Filter Paper: Sheets

Used in schools and as bench protective paper. Fisherbrand Grade 600 Cellulose General Purpose Filter Paper is economical.

Brand:  Fisherbrand™

Code : A0

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11784756

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Used in hospitals, research laboratories, schools, chemical industries, pharmaceutical, cosmetic, food industries
Specifications

Specifications

Sheet
10 to 20 μm
Economical filter Paper used in schools and as bench protective Paper
100/Pk.
260 x 210mm
210 mm
Cellulose
10 to 20 μm
600
0.15 mm
260 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers