missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Grade 480 Silicon Impregnated Phase Separator Papers

Product Code. 10223241 Shop All Sartorius Products
Change view
Click to view available options
Diameter (Metric):
90 mm
110 mm
185 mm
Unit Size:
Pack of 100
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
10223241 90 mm Pack of 100
10049650 110 mm Pack of 100
10039520 185 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10223241 Supplier Sartorius™ Supplier No. FT3602090

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Impregnated with stabilized silicon makes it hydrophobic. Sartorius™ Grade 480 Silicon Impregnated Phase Separator Papers eliminates the need to use separating funnels.

  • Retains water allowing solvents to flow through
  • Flow stops automatically when the entire solvent has passed through

Applications:

  • Filtration of organic solvents that have been contaminated with water to separate the aqueous from the organic phase
  • Filtration of extracting solvents in clinical or medical labs
  • Separation of emulsions that are formed during the extraction of aqueous plant or drug solutions
TRUSTED_SUSTAINABILITY

Specifications

Format Disc
Material Silicon-impregnated Paper
For Use With (Application) Special paper separates aqueous from organic phases
Grade 480
Quantity 100/Pk.
Thickness (Metric) 0.19 mm
Type Qualitative Filter Paper
Diameter (Metric) 90 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.