missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Grade 3236 Seed Test Paper
Click to view available options
Dimensions (L x W):
112 x 20 mm
Unit Size:
Pack of 500
Description
- Provides reliable, reproducible results
- Made from pure cellulose without additives
- Do not contain substances that could influence growth of seeds
- Constant water absorption of paper ensure continuous provision of required amount of water
- Contrast of paper makes evaluation of seeds easier
Specifications
Specifications
| Color | Gray |
| Material | Cellulose |
| For Use With (Application) | For testing medium/large coated seeds (sugar beet, fodder beet, grain, sunflower, rapeseed, mustard) |
| Grade | Grade 3236 |
| Quantity | 1000 Pk. |
| Shape | Pleated strips |
| Type | Seed Test Paper |
| Dimensions (L x W) | 112 x 20 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction