Learn More
Grace™ Reveleris™ Silica Flash Cartridges
Product Code. 12985911
Click to view available options
Bed Weight (Metric):
4 g
Type:
Reveleris SRC Silica Flash Cartridge
Unit Size:
Pack of 20
Product Code. 12985911
Brand: Grace™ 145146130
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Ideal for speed, resolution and capacity
Silica flash cartridges.
  • Withstand higher pressures (up to 200psig) and deliver greater loading capacity
  • Superior, high load silica for excellent resolution
  • Optimally packed to give high plate count
  • Tighter particle size distribution and minimal fines keep backpressure low
  • RFID tag streamlines Reveleris system (sold separately) operation by auto-loading cartridge specifications
  • Compatible with other popular flash systems
Specifications
Material Silica
Type Reveleris SRC Silica Flash Cartridge
Bed Weight (Metric) 4 g
For Use With (Application) For Speed, Resolution and Capacity, Conventional AppliCatIons, Polar Compounds and Popular Flash Systems