missing translation for 'onlineSavingsMsg'
Learn More

GRA Ex-25, MedChemExpress

Product Code. 16430548
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16430548 5 mg 5mg
16490346 2 mg 2mg
16488078 10 mg 10mg
16487846 100 mg 100mg
16473997 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16430548 Supplier Medchem Express Supplier No. HY50675

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GRA Ex-25 is an inhibitor of glucagon receptor, with IC50 of 56 and 55 nM for rat and human glucagon receptors, respectively.

GRA Ex-25 is an inhibitor of glucagon receptor, with IC50 of 56 and 55 nM for rat and human glucagon receptors, respectively.

Specifications

Chemical Name or Material GRA Ex-25
CAS 307983-31-9
Color White
Purity Grade Notes Research
Molecular Formula C29H36F3N3O5
Quantity 5 mg
Solubility Information DMSO : ≥ 32 mg/mL (56.78 mM)
SMILES O=C(O)CCNC(C1=CC=C(CN([C@H]2CC[C@H](C(C)(C)C)CC2)C(NC3=CC=C(OC(F)(F)F)C=C3)=O)C=C1)=O
Molecular Weight (g/mol) 563.61
Formula Weight 563.61
Percent Purity 98.09%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.