missing translation for 'onlineSavingsMsg'
Learn More

GR79236, MedChemExpress

Product Code. 16497668
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16497668

Brand: Medchem Express HY18978

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GR79236 is a highly potent, selective and orally active adenosine A1 receptor agonist with a Kis of 3.1 nM and 1300 nM for A1 and A2 receptors, respectively. GR79236 has anti-nociceptive and anti-inflammatory actions.

GR79236 is a highly potent, selective and orally active adenosine A1 receptor agonist with a Kis of 3.1 nM and 1300 nM for A1 and A2 receptors, respectively. GR79236 has anti-nociceptive and anti-inflammatory actions.

Specifications

Chemical Name or Material GR79236
CAS 124555-18-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C15H21N5O5
Quantity 10 mg
Solubility Information H2O : 100 mg/mL (284.61 mM; Need ultrasonic) ∣DMSO : ≥ 100 mg/mL (284.61 mM)
SMILES OC[C@@H]1[C@H]([C@H]([C@H](N2C=NC3=C2N=CN=C3N[C@@H]4[C@@H](O)CCC4)O1)O)O
Molecular Weight (g/mol) 351.36
Formula Weight 351.36
Percent Purity 99.75%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.