missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ GR 144053 trihydrochloride

Product Code. 18074032
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18074032 10 mg 10mg
18004032 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18074032 Supplier Tocris Bioscience™ Supplier No. 1263/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Glycoprotein IIb/IIIa (integrin αIIbβ3) receptor antagonist. Antithrombotic

A potent and selective platelet fibrinogen receptor glycoprotein IIb/IIIa (GpIIb/IIIa) antagonist (IC50 = 37 nM). Orally active and highly effective at inhibiting thrombus formation in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1215333-48-4
Molecular Formula C18H30Cl3N5O2

Specifications

Quantity 10 mg
Formula Weight 454.83
Purity >99%
Molecular Weight (g/mol) 454.821
Chemical Name or Material GR 144053 trihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.