missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GR 125487 sulfamate, Tocris Bioscience™
Potent and selective 5-HT4 antagonist; active in vivo
£209.16 - £797.91
Chemical Identifiers
CAS | 859502-43-5 |
---|---|
Molecular Formula | C19H29FN4O8S2 |
Molecular Weight (g/mol) | 524.579 |
InChI Key | VQJFDBXITIOGJM-UHFFFAOYSA-N |
Synonym | gr 125487 sulfamate, gr 125487 sulfamate salt, 5-fluoro-2-methoxy-1-2-methylsulfonyl amino ethyl-4-piperidinyl-1h-indole-3-methylcarboxylate sulfamate, tocris-1658, gr-125487 sulfamate salt, 1-2-methanesulfonamido ethyl piperidin-4-yl methyl 5-fluoro-2-methoxy-1h-indole-3-carboxylate; sulfamic acid, 1-2-methylsulfonyl amino ethyl-4-piperidinyl methyl 5-fluoro-2-methoxy-1h-indole-3-carboxylate, 1h-indole-3-carboxylic acid, 5-fluoro-2-methoxy-, 1-2-methylsulfonyl amino ethyl-4-piperidinyl methyl ester, sulfamate 1:1 |
PubChem CID | 6604955 |
IUPAC Name | [1-[2-(methanesulfonamido)ethyl]piperidin-4-yl]methyl 5-fluoro-2-methoxy-1H-indole-3-carboxylate;sulfamic acid |
SMILES | COC1=C(C2=C(N1)C=CC(=C2)F)C(=O)OCC3CCN(CC3)CCNS(=O)(=O)C.NS(=O)(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18065422
|
Tocris Bioscience
1658/10 |
10 mg |
£209.16
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18055002
|
Tocris Bioscience
1658/50 |
50 mg |
£797.91
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potent and selective 5-HT4 receptor antagonist (Ki = 0.19 nM for porcine 5-HT4). Centrally active following systemic administration in vivo.Chemical Identifiers
859502-43-5 | |
524.579 | |
gr 125487 sulfamate, gr 125487 sulfamate salt, 5-fluoro-2-methoxy-1-2-methylsulfonyl amino ethyl-4-piperidinyl-1h-indole-3-methylcarboxylate sulfamate, tocris-1658, gr-125487 sulfamate salt, 1-2-methanesulfonamido ethyl piperidin-4-yl methyl 5-fluoro-2-methoxy-1h-indole-3-carboxylate; sulfamic acid, 1-2-methylsulfonyl amino ethyl-4-piperidinyl methyl 5-fluoro-2-methoxy-1h-indole-3-carboxylate, 1h-indole-3-carboxylic acid, 5-fluoro-2-methoxy-, 1-2-methylsulfonyl amino ethyl-4-piperidinyl methyl ester, sulfamate 1:1 | |
[1-[2-(methanesulfonamido)ethyl]piperidin-4-yl]methyl 5-fluoro-2-methoxy-1H-indole-3-carboxylate;sulfamic acid |
C19H29FN4O8S2 | |
VQJFDBXITIOGJM-UHFFFAOYSA-N | |
6604955 | |
COC1=C(C2=C(N1)C=CC(=C2)F)C(=O)OCC3CCN(CC3)CCNS(=O)(=O)C.NS(=O)(=O)O |
Specifications
859502-43-5 | |
gr 125487 sulfamate, gr 125487 sulfamate salt, 5-fluoro-2-methoxy-1-2-methylsulfonyl amino ethyl-4-piperidinyl-1h-indole-3-methylcarboxylate sulfamate, tocris-1658, gr-125487 sulfamate salt, 1-2-methanesulfonamido ethyl piperidin-4-yl methyl 5-fluoro-2-methoxy-1h-indole-3-carboxylate; sulfamic acid, 1-2-methylsulfonyl amino ethyl-4-piperidinyl methyl 5-fluoro-2-methoxy-1h-indole-3-carboxylate, 1h-indole-3-carboxylic acid, 5-fluoro-2-methoxy-, 1-2-methylsulfonyl amino ethyl-4-piperidinyl methyl ester, sulfamate 1:1 | |
COC1=C(C2=C(N1)C=CC(=C2)F)C(=O)OCC3CCN(CC3)CCNS(=O)(=O)C.NS(=O)(=O)O | |
524.579 | |
524.58 | |
GR 125487 sulfamate |
C19H29FN4O8S2 | |
VQJFDBXITIOGJM-UHFFFAOYSA-N | |
[1-[2-(methanesulfonamido)ethyl]piperidin-4-yl]methyl 5-fluoro-2-methoxy-1H-indole-3-carboxylate;sulfamic acid | |
6604955 | |
>98% |
Safety and Handling
Recommended Storage : Desiccate at Room Temperature