missing translation for 'onlineSavingsMsg'
Learn More

Gr-1/Ly-6G, Rabbit anti-Mouse, Clone: 1A8, Novus Biologicals™

Product Code. 18350160 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
Unit Size:
0.2mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18350160 0.2 mg 0.2mg
18208538 0.025 mg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18350160 Supplier Novus Biologicals Supplier No. NBP2810530.2MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Gr-1/Ly-6G Monoclonal specifically detects Gr-1/Ly-6G in Mouse samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Gr-1/Ly-6G
Applications Flow Cytometry
Classification Monoclonal
Clone 1A8
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry
Formulation PBS
Gene Alias C6orf23, G6D, LY6-D, LY6G6D, lymphocyte antigen 6 family member G6D, MEGT1, NG25
Gene Symbols LY6G6D
Host Species Rabbit
Immunogen LEW/N rats immunized with 1.5x10E8 Ly-6G-transfected EL4J cells
Purification Method Protein A purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 58530
Test Specificity Recognises the murine GPI-anchored protein Ly6G, a marker of granulocyte differentiation, maturation and, together with Ly6C, a component of the Granulocyte Receptor-1 Antigen (Gr-1). Unlike another commonly used clone, RB6-8C5, 1A8 does not bind Ly6C.
Target Species Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.