missing translation for 'onlineSavingsMsg'
Learn More

Gr-1 (Ly-6G/Ly-6C) Rat, anti-Mouse, Biotin, Clone: RB6-8C5, STEMCELL Technologies

Rat Monoclonal Antibody

£298.35 - £311.85

Specifications

Antigen Gr-1 (Ly-6G/Ly-6C)
Clone RB6-8C5
Applications Immunoprecipitation, Western Blot, Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Conjugate Biotin
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17168601
Encompass_Preferred
View Documents
Stemcell Technologies
60028BT.1
25 μg
£311.85
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17158601
Encompass_Preferred
View Documents
Stemcell Technologies
60028BT
100 μg
£298.35
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • The RB6-8C5 antibody reacts with the structurally related murine Ly-6G (21 - 25 kDa) and Ly-6C (14 - 16 kDa) GPI-anchored proteins, which together comprise the granulocyte receptor-1 antigen (Gr-1)
  • Gr-1 is expressed on monocytes, neutrophils, subsets of macrophages, plasmacytoid dendritic cells, and T cells
  • Monocytes in the bone marrow transiently express Gr-1 during development and the expression level is strongly correlated with granulocyte differentiation and maturation
  • In the periphery Gr-1 is found predominantly on neutrophils and is a useful marker for these cells
  • Whereas the RB6-8C5 binds to both Ly-6G and Ly-6C, another commonly used anti-Gr-1 antibody, clone 1A8, binds specifically to Ly-6G
  • This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse Neutrophil Enrichment Kit (Catalog #19762)
Specifications

Specifications

Gr-1 (Ly-6G/Ly-6C)
Immunoprecipitation, Western Blot, Flow Cytometry, Immunocytochemistry
Biotin
Mouse
Primary
RB6-8C5
Monoclonal
Rat
IgG2b κ
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.