missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gr-1/Ly-6G Antibody (RB6-8C5) [Janelia Fluor™ 635], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB1037JF635
This item is not returnable.
View return policy
Description
Gr-1/Ly-6G Monoclonal antibody specifically detects Gr-1/Ly-6G in Mouse samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunoprecipitation,Immunocytochemistry,CyTOF
Specifications
| Gr-1/Ly-6G | |
| Monoclonal | |
| Janelia Fluor 635 | |
| C6orf23, G6D, LY6-D, LY6G6D, lymphocyte antigen 6 family member G6D, MEGT1, NG25 | |
| The immunogen for this antibody is Ly-6G (Gr-1, Gr1). | |
| RUO | |
| 58530 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry, CyTOF | |
| RB6-8C5 | |
| 50mM Sodium Borate | |
| Rat | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction