missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gr-1/Ly-6G Antibody (AP-MAB0866), Alexa Fluor™ 532, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-06691AF532
This item is not returnable.
View return policy
Description
Gr-1/Ly-6G Monoclonal antibody specifically detects Gr-1/Ly-6G in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Gr-1/Ly-6G | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| AP-MAB0866 | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| C6orf23, G6D, LY6-D, LY6G6D, lymphocyte antigen 6 family member G6D, MEGT1, NG25 | |
| Mouse spontaneous T lymphoma line TK1 | |
| 0.1 mL | |
| Innate Immunity | |
| 58530 | |
| Store at 4°C in the dark. | |
| IgG2 |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion