missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Gr-1/Ly-6G Antibody (3D6E10), Novus Biologicals™
Mouse Monoclonal Antibody
£236.00 - £420.00
Specifications
| Antigen | Gr-1/Ly-6G |
|---|---|
| Clone | 3D6E10 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunocytochemistry/ Immunofluorescence 1:200 - 1:1000 |
| Applications | Flow Cytometry, ELISA, Western Blot, Immunocytochemistry/Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30039839
|
Novus Biologicals™
NBP3-27164-0.025ml |
0.025 mL |
£236.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30033054
|
Novus Biologicals™
NBP3-27164-0.1ml |
0.1 mL |
£420.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Gr-1/Ly-6G Monoclonal antibody specifically detects Gr-1/Ly-6G in Human,Monkey,Mouse,Rat samples. It is validated for Flow Cytometry,ELISA,Western Blot,Immunocytochemistry/ ImmunofluorescenceSpecifications
| Gr-1/Ly-6G | |
| 1 mg/mL | |
| Flow Cytometry, ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Innate Immunity | |
| PBS | |
| 58530 | |
| IgG1 | |
| Protein G purified |
| 3D6E10 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunocytochemistry/ Immunofluorescence 1:200 - 1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Monkey, Mouse, Rat | |
| C6orf23, G6D, LY6-D, LY6G6D, lymphocyte antigen 6 family member G6D, MEGT1, NG25 | |
| Purified recombinant fragment of human Gr-1/Ly-6G (AA: 20-104) expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title