missing translation for 'onlineSavingsMsg'
Learn More

GPX8 Antibody (8-1), Novus Biologicals™

Product Code. 18365244 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18365244 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18365244 Supplier Novus Biologicals Supplier No. NBP313659

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GPX8 Monoclonal antibody specifically detects GPX8 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPX8
Applications Western Blot
Classification Monoclonal
Clone 2023-08-01 00:00:00
Conjugate Unconjugated
Formulation PBS
Gene Alias EC 1.11.1, EC 1.11.1.9, EPLA847, glutathione peroxidase 8 (putative), GPx-8, GSHPx-8, probable glutathione peroxidase 8, UNQ847
Host Species Mouse
Immunogen The immunogen used to generate this antibody corresponds to human GPX8.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 493869
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.