missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPX7 Monoclonal Antibody (2704)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA123306
This item is not returnable.
View return policy
Description
Recommended positive controls: HeLa whole cell extract. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Human GPX 7 is believed to be a member of the glutathione peroxidase family of proteins. These proteins play important roles in cellular defense mechanisms against oxidative damage. This is accomplished by catalyzing the reduction of hydroperoxides and hydrogen peroxide through the reducing substrate, glutathione.
Specifications
| GPX7 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 20% glycerol and no preservative; pH 7 | |
| Q96SL4 | |
| GPX7 | |
| Human GPX7. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| 2704 | |
| Unconjugated | |
| GPX7 | |
| 3110050F08Rik; AI327032; CL683; FLJ14777; glutathione peroxidase 6; glutathione peroxidase 7; GPX6; Gpx7; GPx-7; GSHPx-7; HGNC:4559; non-selenocysteine containing phospholipid hydroperoxide glutathione peroxidase; NPGPx; UNQ469/PRO828 | |
| Mouse | |
| Protein G | |
| RUO | |
| 2882 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction