missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPX4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14432-1-AP-20UL
This item is not returnable.
View return policy
Description
Glutathione peroxidase catalyzes the reduction of hydrogen peroxide, organic hydroperoxide, and lipid peroxides by reduced glutathione and functions in the protection of cells against oxidative damage. Human plasma glutathione peroxidase has been shown to be a selenium-containing enzyme and the UGA codon is translated into a selenocysteine. Through alternative splicing and transcription initiation, rat produces proteins that localize to the nucleus, mitochondrion, and cytoplasm. In humans, experimental evidence for alternative splicing exists; alternative transcription initiation and the cleavage sites of the mitochondrial and nuclear transit peptides need to be experimentally verified.Specifications
| GPX4 | |
| Polyclonal | |
| Unconjugated | |
| GPX4 | |
| Glutathione peroxidase 4, GPx 4, GPX4, GSHPx 4, MCSP, PHGPx, snGPx, snPHGPx | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2879, 29328, 625249 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Western Blot, Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P36969, Q91XR8, Q91XR9 | |
| GPX4 | |
| GPX4 Fusion Protein Ag5809 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering