missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPX2 Recombinant Rabbit Monoclonal Antibody (1X5K1)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA555456
This item is not returnable.
View return policy
Description
Immunogen Sequence: QFGHQENCQN EEILNSLKYV RPGGGYQPTF TLVQKCEVNG QNEHPVFAYL KDKLPYPYDD PFSLMTDPKL IIWSPVRRSD VAWNFEKFLI GPEGEPFRRY SRTFP.
This gene is a member of the glutathione peroxidase family and encodes a selenium-dependent glutathione peroxidase that is one of two isoenzymes responsible for the majority of the glutathione-dependent hydrogen peroxide-reducing activity in the epithelium of the gastrointestinal tract. Studies in knockout mice indicate that mRNA expression levels respond to luminal microflora, suggesting a role of the ileal glutathione peroxidases in preventing inflammation in the GI tract.
Specifications
| GPX2 | |
| Recombinant Monoclonal | |
| 2 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P18283 | |
| GPX2 | |
| Synthetic peptide. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 1X5K1 | |
| Unconjugated | |
| GPX2 | |
| gastrointestinal glutathione peroxidase; GI-GPx; Glutathione peroxidase 2; glutathione peroxidase 2 (gastrointestinal); glutathione peroxidase-gastrointestinal; Glutathione peroxidase-related protein 2; GPRP; GPRP-2; GPX2; GPx-2; GPX-GI; GSHPx-2; GSHPX-GI; HGNC:4554; intestinal GPx | |
| Rabbit | |
| Protein A | |
| RUO | |
| 2877 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction