missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GPX2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 11326373
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11326373 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11326373 Supplier Invitrogen™ Supplier No. PA518397

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is predicted to react with bovine, canine, mouse, porcine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.

This gene is a member of the glutathione peroxidase family and encodes a selenium-dependent glutathione peroxidase that is one of two isoenzymes responsible for the majority of the glutathione-dependent hydrogen peroxide-reducing activity in the epithelium of the gastrointestinal tract. Studies in knockout mice indicate that mRNA expression levels respond to luminal microflora, suggesting a role of the ileal glutathione peroxidases in preventing inflammation in the GI tract.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPX2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene GPX2
Gene Accession No. P18283
Gene Alias gastrointestinal glutathione peroxidase; GI-GPx; Glutathione peroxidase 2; glutathione peroxidase 2 (gastrointestinal); glutathione peroxidase-gastrointestinal; Glutathione peroxidase-related protein 2; GPRP; GPRP-2; GPX2; GPx-2; GPX-GI; GSHPx-2; GSHPX-GI; HGNC:4554; intestinal GPx
Gene Symbols GPX2
Host Species Goat
Immunogen Synthetic peptide sequence (SLDGEKVDFN) corresponding to the internal amino acids of GPX2 (aa 15-25).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2877
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.