missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPX1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588073
This item is not returnable.
View return policy
Description
Immunogen sequence: RPGGGFEPNF MLFEKCEVNG AGAHPLFAFL REALPAPSDD ATALMTDPKL ITWSPVCRND VAWNFEKFLV GPDGVPLRRY SRRFQTIDIE PDIEALLSQG P Positive Samples: 293T, Mouse liver, Mouse kidney, Rat liver, Rat spleen; Cellular Location: Cytoplasm.
Glutathione peroxidases (Gpxs) are ubiquitously expressed proteins which catalyze the reduction of hydrogen peroxides and organic hydroperoxides by glutathione. There are several isoforms which differ in their primary structure and localization. The classical cytosolic/mitochondrial GPx1 (cGPx) is a selenium-dependent enzyme, first of the GPx family to be discovered. GPx2, also known as gastrointestinal GPx (GI-GPx), is an intracellular enzyme expressed only at the epithelium of the gastrointestinal tract. Extracellular plasma GPx (pGPx or GPx3) is mainly expressed by the kidney from where it is released into the blood circulation. Phospholipid hydroperoxide GPx4 (PH-GPx) expressed in most tissues, can reduce many hydroperoxides including hydroperoxides integrated in membranes, hydroperoxy lipids in low density lipoprotein or thymine. All mammalian GPx family members, except for the recently described Cys containing GPx3 and epididymis-specific secretory GPx (eGPx or GPx5) isoforms, possess selenocysteine at the active site.
Specifications
| GPX1 | |
| Polyclonal | |
| Unconjugated | |
| GPX1 | |
| AI195024; AL033363; Cellular glutathione peroxidase; CGPx; cytosolic glutathione peroxidase; glutathione peroxidase; glutathione peroxidase 1; Gpx; Gpx1; GPx-1; GPXD; GSHPx; GSHPX1; GSHPx-1; HGNC:4553; MGC14399; MGC88245; OTTHUMP00000210766; Selenium-dependent glutathione peroxidase 1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 14775, 24404, 2876 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 3.55 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P04041, P07203, P11352 | |
| GPX1 | |
| A synthetic peptide corresponding to a sequence within amino acids 100-200 of human GPX1 (NP_0005722). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction