missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GPT2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15730222 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15730222 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15730222 Supplier Invitrogen™ Supplier No. PA562426

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: PVSGQAAMDI VVNPPVAGEE SFEQFSREKE SVLGNLA Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 95%.

GPT and GPT2 (EC 2.6.1.2), also known as alanine transaminases, are pyridoxal enzymes that catalyze the reversible transamination between alanine and 2-oxoglutarate to form pyruvate and glutamate. By mediating the conversion of these 4 major intermediate metabolites, these transaminases have roles in gluconeogenesis and in amino acid metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPT2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GPT2
Gene Accession No. Q8TD30
Gene Alias 4631422C05Rik; AAT2; Alanine aminotransferase 2; ALT2; AU014768; AU041193; C87201; Cc2-5; glutamate pyruvate transaminase 2; glutamic pyruvate transaminase (alanine aminotransferase) 2; glutamic pyruvate transaminase 2; glutamic--alanine transaminase 2; glutamic-pyruvate transaminase 2; Glutamic--pyruvic transaminase 2; GPT 2; GPT2; im:7150662; LOW QUALITY PROTEIN: alanine aminotransferase 2; MRT49; sb:cb580; zgc:165657
Gene Symbols GPT2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GPT2. Recombinant protein control fragment (Product #RP-101440).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84706
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.