missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPT Monoclonal Antibody (C4), Invitrogen™
Mouse Monoclonal Antibody
£161.00 - £350.00
Specifications
| Antigen | GPT |
|---|---|
| Clone | C4 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17920173
|
Invitrogen™
MA550914 |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17980643
|
Invitrogen™
MA550913 |
200 μL |
£350.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GPT(Glutamate-pyruvate transaminase), also known as alanine aminotransferase, catalyzes the reversible conversion of L-alanine and alpha-ketoglutarate to L-glutamate and pyruvate. This protein has 2 distinct molecular and genetic forms: one cytoplasmic (soluble) (GPT1) and one mitochondrial (GPT2). See ALTQTL1 and ALTQTL2 for information on quantitative trait loci influencing the plasma level of alanine aminotransferase.Specifications
| GPT | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| 1300007J06Rik; 2310022B03Rik; AAT1; alanine aminotransferase; alanine aminotransferase 1; ALT; ALT1; glutamate pyruvate transaminase 1; glutamic pyruvate transaminase; glutamic pyruvic transaminase (alanine aminotrasferase); glutamic pyruvic transaminase 1, soluble; glutamic pyruvic transaminase, soluble; glutamic-alanine transaminase 1; glutamic--alanine transaminase 1; glutamic-pyruvate transaminase (alanine aminotransferase); glutamic--pyruvic transaminase; glutamic--pyruvic transaminase 1; GPT; GPT 1; Gpt1; Gpt-1 | |
| GPT | |
| IgG1 κ | |
| Protein A/G | |
| GPT |
| C4 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Rat | |
| P24298, P25409 | |
| 2875, 81670 | |
| Recombinant Human ALT protein, Ile255-Thr492 + LEHHHHHH (Accession #P24298). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title