missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPT Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-31525
This item is not returnable.
View return policy
Description
GPT Polyclonal specifically detects GPT in Human, Mouse samples. It is validated for Western Blot.
Specifications
| GPT | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| GPT | |
| Recombinant protein encompassing a sequence within the N-terminus region of human GPT. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Tested in human. Predicted cross-reactivity based on sequence identity: Bovine (89%). Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.47mg/mL | |
| Western Blot 1:500-1:3000 | |
| AAT1alanine aminotransferase 1, ALT1, ALT1EC 2.6.1.2, Glutamate pyruvate transaminase 1, glutamic-alanine transaminase 1, Glutamic--alanine transaminase 1, glutamic-pyruvate transaminase (alanine aminotransferase), Glutamic--pyruvic transaminase 1, GPT1GPT 1 | |
| Rabbit | |
| 55 kDa | |
| 0.1 mL | |
| metabolism | |
| 2875 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction