missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPT Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38016-100ul
This item is not returnable.
View return policy
Description
GPT Polyclonal antibody specifically detects GPT in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| GPT | |
| Polyclonal | |
| Western Blot 1:200 - 1:1000, ELISA | |
| AAT1alanine aminotransferase 1, ALT1, ALT1EC 2.6.1.2, Glutamate pyruvate transaminase 1, glutamic-alanine transaminase 1, Glutamic--alanine transaminase 1, glutamic-pyruvate transaminase (alanine aminotransferase), Glutamic--pyruvic transaminase 1, GPT1GPT 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-190 of human GPT (NP_005300.1).,, Sequence:, MASSTGDRSQAVRHGLRAKVLTLDGMNPRVRRVEYAVRGPIVQRALELEQELRQGVKKPFTEVIRANIGDAQAMGQRPITFLRQVLALCVNPDLLSSPNFPDDAKKRAERILQACGGHSLGAYSVSSGIQLIREDVARYIERRDGGIPADPNNVFLSTGASDAIVTVLKLLVAGEGHTRTGVLIPIPQYP | |
| 100 μL | |
| metabolism | |
| 2875 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction