missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPSN2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563311
This item is not returnable.
View return policy
Description
Immunogen sequence: MKHYEVEILD AKTREKLCFL DKVEPHATIA EIKNLF Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.
Microsomal long and very long chain fatty acid elongation uses malonyl-CoA as the 2-carbon donor and consists of 4 sequential reactions. GPSN2 catalyzes the final step, reducing trans-2, 3-enoyl-CoA to saturated acyl-CoA. Microsomal long and very long chain fatty acid elongation uses malonyl-CoA as the 2-carbon donor and consists of 4 sequential reactions. TER catalyzes the final step, reducing trans-2, 3-enoyl-CoA to saturated acyl-CoA (Moon and Horton, 2003 [PubMed 12482854]).[supplied by OMIM].
Specifications
| GPSN2 | |
| Polyclonal | |
| Unconjugated | |
| TECR | |
| 2410016D23Rik; A230102P12Rik; AI173355; D17Ertd178e; glycoprotein, synaptic 2; GPSN2; hypothetical protein LOC614105; MRT14; neuroprotective protein 13; SC2; Synaptic glycoprotein SC2; Tecr; TER; trans-2,3-enoyl-CoA reductase; trans-2,3-enoyl-CoA reductase; very-long-chain enoyl-CoA reductase; very-long-chain enoyl-CoA reductase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9524 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NZ01 | |
| TECR | |
| Recombinant protein corresponding to Human GPSN2. Recombinant protein control fragment (Product #RP-104286). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction