missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPSM3 Antibody (1F11), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00063940-M01
This item is not returnable.
View return policy
Description
GPSM3 Monoclonal antibody specifically detects GPSM3 in Human samples. It is validated for Western Blot, ELISA
Specifications
| GPSM3 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| Activator of G-protein signaling 4, AGS4C6orf9, G18.1bC. elegans), G18.2, G18chromosome 6 open reading frame 9, G-protein signaling modulator 3, G-protein signalling modulator 3 (AGS3-like, C. elegans), G-protein-signaling modulator 3, NG1, Protein G18 | |
| GPSM3 (AAH18724, 1 a.a. ∽ 160 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MEAERPQEEEDGEQGPPQDEEGWPPPNSTTRPWRSAPPSPPPPGTRHTALGPRSASLLSLQTELLLDLVAEAQSRRLEEQRATFYTPQNPSSLAPAPLRPLEDREQLYSTILSHQCQRMEAQRSEPPLPPGGQELLELLLRVQGGGRMEEQRSRPPTHTC | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 1F11 | |
| Western Blot, ELISA | |
| AAH18724 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 63940 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction