missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPSM2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26798-1-AP-150UL
This item is not returnable.
View return policy
Description
The Tousled-like kinases, first described in Arabadopsis, are nuclear serine/threonine kinases that are potentially involved in the regulation of chromatin assembly.[supplied by OMIM]Specifications
| GPSM2 | |
| Polyclonal | |
| Unconjugated | |
| GPSM2 | |
| GPSM2, LGN, Mosaic protein LGN, Pins | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 29899, 362021, 76123 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P81274, Q8VDU0 | |
| Gpsm2 | |
| GPSM2 Fusion Protein Ag25195 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction