missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPS2 Recombinant Rabbit Monoclonal Antibody (HL2276)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA550471
This item is not returnable.
View return policy
Description
GPS2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes a protein involved in G protein-mitogen-activated protein kinase (MAPK) signaling cascades. When overexpressed in mammalian cells, this gene could potently suppress a RAS- and MAPK-mediated signal and interfere with JNK activity, suggesting that the function of this gene may be signal repression. The encoded protein is an integral subunit of the NCOR1-HDAC3 (nuclear receptor corepressor 1-histone deacetylase 3) complex, and it was shown that the complex inhibits JNK activation through this subunit and thus could potentially provide an alternative mechanism for hormone-mediated antagonism of AP1 (activator protein 1) function.
Specifications
| GPS2 | |
| Recombinant Monoclonal | |
| 2 mg/mL | |
| PBS with no preservative | |
| Q13227 | |
| GPS2 | |
| Recombinant fragmemt of human GPS2. | |
| 100 μL | |
| Primary | |
| Human, Zebrafish | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| HL2276 | |
| Unconjugated | |
| GPS2 | |
| AI505953; AMF-1; G protein pathway suppressor 2; GPS2; GPS-2 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 2874, 393948 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction