missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPRC5D Antibody (19), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-26101MFV610
This item is not returnable.
View return policy
Description
GPRC5D Monoclonal antibody specifically detects GPRC5D in Human samples. It is validated for Flow Cytometry
Specifications
| GPRC5D | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 19 | |
| Flow Cytometry | |
| G protein-coupled receptor, family C, group 5, member D, G-protein coupled receptor family C group 5 member D, MGC129713, MGC129714, orphan G-protein coupled receptor | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human GPRC5D. | |
| 0.1 mL | |
| GPCR | |
| 55507 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction