Learn More
Invitrogen™ GPRC5C Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA532856
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Orangutan, Gibbon, Monkey, Marmoset, Dog, Bat, Bovine, Elephant, Horse (100%); Mouse, Rat, Hamster, Rabbit, Opossum, Turkey, Chicken, Platypus, Lizard (95%); Panda (84%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
This polyclonal antibody was raised in Rabbit against GPRC5C. This antibody is unconjugated. This antibody is useful for testing in the following assays: IHC-P. The specificity of this antibody has been described as follows: Human GPRC5C. BLAST analysis of the peptide immunogen showed no homology with other human proteins.
Specifications
| GPRC5C | |
| Polyclonal | |
| Unconjugated | |
| GPRC5C | |
| 1110028I06Rik; 3200002M13Rik; G protein-coupled receptor class C group 5 member C; G protein-coupled receptor, class C, group 5, member C; G protein-coupled receptor, family C, group 5, member C; Gprc5c; G-protein coupled receptor family C group 5 member C; MGC131820; orphan G-protein coupled receptor; PSEC0087; RAIG3; RAIG-3; retinoic acid inducible protein 3; retinoic acid responsive gene protein; retinoic acid-induced gene 3 protein | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100060724, 483288, 535664, 55890 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q2YDG0, Q9NQ84 | |
| GPRC5C | |
| Synthetic 19 amino acid peptide from C-terminal cytoplasmic domain of human GPRC5C. | |
| 50 μg | |
| Primary | |
| Human, Canine, Monkey, Bovine, Horse, Bat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.