missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPRC5C Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00055890-B01P
This item is not returnable.
View return policy
Description
GPRC5C Polyclonal antibody specifically detects GPRC5C in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence
Specifications
| GPRC5C | |
| Polyclonal | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunocytochemistry/ Immunofluorescence 1:10-1:500 | |
| NP_061123.3 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunocytochemistry | |
| Unconjugated | |
| PBS (pH 7.4) | |
| G protein-coupled receptor, family C, group 5, member C, MGC131820, RAIG-3G-protein coupled receptor family C group 5 member C, RAIG3orphan G-protein coupled receptor, retinoic acid responsive gene protein, Retinoic acid-induced gene 3 protein | |
| GPRC5C (NP_061123.3, 1 a.a. - 453 a.a.) full-length human protein. MGTQPEPGLGARMAIHKALVMCLGLPLFLFPGAWAQGHVPPGCSQGLNPLYYNLCDRSGAWGIVLEAVAGAGIVTTFVLTIILVASLPFVQDTKKRSLLGTQVFFLLGTLGLFCLVFACVVKPDFSTCASRRFLFGVLFAICFSCLAAHVFALNFLARKNHGPRGWVIFTVALLLTLVEVIINTEWLIITLVRGSGEGGPQGNSSAGWAVASPCAIANMDFVMALIYVMLLLLGAFLGAWPALCGRYKRWRKHGVFVLLTTATSVAIWVVWIVMYTYGNKQHNSPTWDDPTLAIALAANAWAFVLFYVIPEVSQVTKSSPEQSYQGDMYPTRGVGYETILKEQKGQSMFVENKAFSMDEPVAAKRPVSPYSGYNGQLLTSVYQPTEMALMHKVPSEGAYDIILPRATANSQVMGSANSTLRAEDMYSAQSHQAATPPKDGKNSQVFRNPYVWD | |
| 0.05 mg | |
| GPCR | |
| 55890 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction