missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GPRC5B Recombinant Rabbit Monoclonal Antibody (K01_1R93)

Product Code. 30830424
Change view
Click to view available options
Quantity:
50 μL
100 μL
Unit Size:
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30830424 50 μL 50µL
30829198 100 μL -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30830424 Supplier Invitrogen™ Supplier No. MA564904
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

GPRC5B Recombinant Monoclonal Antibody for Western Blot

This gene encodes a member of the type 3 G protein-coupled receptor family. Members of this superfamily are characterized by a signature 7-transmembrane domain motif. The encoded protein may modulate insulin secretion and increased protein expression is associated with type 2 diabetes. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPRC5B
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1R93
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene GPRC5B
Gene Accession No. Q923Z0, Q9NZH0
Gene Alias A-69G12.1; G protein-coupled receptor class C group 5 member B; G protein-coupled receptor, class C, group 5, member B; G protein-coupled receptor, family C, group 1, member B; GPRC5B; G-protein coupled receptor family C group 5 member B; RAIG2; RAIG-2; retinoic acid responsive gene protein; retinoic acid-induced gene 2 protein
Gene Symbols GPRC5B
Host Species Rabbit
Immunogen A synthetic peptide of human GPRC5B.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 293546, 51704, 64297
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.