Learn More
Invitrogen™ GPRC5B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA532854
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Orangutan, Monkey, Marmoset, Dog, Bovine, Elephant, Panda, Horse, Pig (100%); Gibbon, Opossum, Turkey, Chicken (94%); Xenopus (89%); Rat (83%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
This gene encodes a member of the type 3 G protein-coupled receptor family. Members of this superfamily are characterized by a signature 7-transmembrane domain motif. The encoded protein may modulate insulin secretion and increased protein expression is associated with type 2 diabetes. Alternative splicing results in multiple transcript variants.
Specifications
| GPRC5B | |
| Polyclonal | |
| Unconjugated | |
| GPRC5B | |
| A-69G12.1; G protein-coupled receptor class C group 5 member B; G protein-coupled receptor, class C, group 5, member B; G protein-coupled receptor, family C, group 1, member B; GPRC5B; G-protein coupled receptor family C group 5 member B; RAIG2; RAIG-2; retinoic acid responsive gene protein; retinoic acid-induced gene 2 protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100052112, 100623340, 100856475, 516334, 51704 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q9NZH0 | |
| GPRC5B | |
| Synthetic 18 amino acid peptide from C-terminal cytoplasmic domain of human GPRC5B. | |
| 50 μg | |
| Primary | |
| Human, Canine, Monkey, Bovine, Pig, Horse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.