missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
GPRC5B Monoclonal antibody specifically detects GPRC5B in Human samples. It is validated for Flow Cytometry,CyTOF
Specifications
Specifications
| Antigen | GPRC5B |
| Applications | Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Clone | 575926 |
| Conjugate | Janelia Fluor 585 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | G protein-coupled receptor, family C, group 1, member B, G protein-coupled receptor, family C, group 5, member B, RAIG-2A-69G12.1, RAIG2G-protein coupled receptor family C group 5 member B, retinoic acid responsive gene protein, Retinoic acid-induced gene 2 protein |
| Host Species | Mouse |
| Immunogen | NS0 mouse myeloma cell line transfected with human GPRC5B, Accession # Q9NZH0 |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?