missing translation for 'onlineSavingsMsg'
Learn More

GPRC5B Antibody (575926), CoraFluor™ 1, Novus Biologicals™

Product Code. 30152668 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30152668 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30152668 Supplier Novus Biologicals Supplier No. FAB10253CL1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GPRC5B Monoclonal antibody specifically detects GPRC5B in Human samples. It is validated for Flow Cytometry,CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPRC5B
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 575926
Conjugate CoraFluor 1
Formulation PBS
Gene Alias G protein-coupled receptor, family C, group 1, member B, G protein-coupled receptor, family C, group 5, member B, RAIG-2A-69G12.1, RAIG2G-protein coupled receptor family C group 5 member B, retinoic acid responsive gene protein, Retinoic acid-induced gene 2 protein
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human GPRC5B, Accession # Q9NZH0
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 51704
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.